2-(Bromomethyl)-2-phenyl-1,3-dioxolane ≥95%
Supplier: Matrix Scientific
042587-1G
042587-500MG
042587-5G
101856-484EA
278.46
USD
101856-484
101856-486
101856-488
2-(Bromomethyl)-2-phenyl-1,3-dioxolane ≥95%
2-(Bromomethyl)-2-phenyl-1,3-dioxolane
Hazard: Irritant
Formula:
C₁₀H₁₁BrO₂ Melting Pt: 63…65 °C |
MDL Number:
MFCD00225209 CAS Number: 3418-21-1 |
Specification Test Results
Melting point | 63-65°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...