5-(3-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde ≥95%
Supplier: Matrix Scientific
042528-500MG
042528-1G
042528-5G
101856-202EA
747.69
USD
101856-202
101856-204
101856-206
5-(3-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde ≥95%
5-(3-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Hazard: Irritant
Formula:
C₁₂H₈ClF₃N₂O₂ MW: 304.66 g/mol Melting Pt: 61…63 °C |
MDL Number:
MFCD00243688 CAS Number: 282523-36-8 |
Specification Test Results
Melting point | 61-63°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...