1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carbaldehyde ≥95%
Supplier: Matrix Scientific
042527-5G
042527-1G
042527-500MG
101856-196EA
747.69
USD
101856-196
101856-198
101856-200
1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carbaldehyde ≥95%
1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carbaldehyde
Hazard: Irritant
Formula:
C₁₃H₈F₆N₂O₂ MW: 338.21 g/mol Melting Pt: 81…82 °C |
MDL Number:
MFCD00243685 CAS Number: 158712-22-2 |
Specification Test Results
Melting point | 81-82°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...