Ethyl-2,6-dimethylimidazo[1,2-a]pyridine-3-carboxylate ≥95%
Supplier: Matrix Scientific
042512-1G
042512-500MG
101856-120EA
184.62
USD
101856-120
101856-122
Ethyl-2,6-dimethylimidazo[1,2-a]pyridine-3-carboxylate ≥95%
Ethyl 2,6-dimethylimidazo[1,2-a]pyridine-3-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₄N₂O₂ Melting Pt: 70…72 °C |
CAS Number:
81438-51-9 |
Specification Test Results
Melting point | 70-72°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...