Ethyl-2,8-dimethylimidazo[1,2-a]pyridine-3-carboxylate ≥95%
Supplier: Matrix Scientific
042511-5G
042511-1G
042511-500MG
101856-114EA
523.25
USD
101856-114
101856-116
101856-118
Ethyl-2,8-dimethylimidazo[1,2-a]pyridine-3-carboxylate ≥95%
Ethyl 2,8-dimethylimidazo[1,2-a]pyridine-3-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₄N₂O₂ Melting Pt: 79…82 °C |
MDL Number:
MFCD00139511 CAS Number: 241146-66-7 |
Specification Test Results
Melting point | 79-82°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...