1-Oxo-1,2-dihydro-4-isoquinolinecarboxylic acid ≥95%

Supplier: Matrix Scientific

042502-500MG 042502-1G 042502-5G
101856-066EA 281.54 USD
101856-066 101856-068 101856-070
1-Oxo-1,2-dihydro-4-isoquinolinecarboxylic acid ≥95%
1-Oxo-1,2-dihydro-4-isoquinolinecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₇NO₃
Melting Pt: 310 °C
MDL Number: MFCD00140316
CAS Number: 34014-51-2

Order Now

Specification Test Results

Melting point 310°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR