4-Carboxy-3-fluorophenylboronic acid

Supplier: Matrix Scientific

Synonyms: 4-Carboxy-3-fluorobenzeneboronic acid4-Borono-2-fluorobenzoic acid

035568-25G 035568-5G
101839-516EA 44.62 USD
101839-516 101839-518
4-Carboxy-3-fluorophenylboronic acid
4-Carboxy-3-fluorophenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₇H₆BFO₄
MW: 183.93 g/mol
Melting Pt: 236…240 °C
MDL Number: MFCD01114671
CAS Number: 120153-08-4

Order Now

Specification Test Results

Melting point 236-240°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR