4-Butylphenylboronic acid

Supplier: Matrix Scientific

Synonyms: 4-n-Butylbenzeneboronic acid

035563-5G
101839-510EA 38.46 USD
101839-510
4-Butylphenylboronic acid
4-Butylphenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₁₀H₁₅BO₂
MW: 178.04 g/mol
Melting Pt: 91…97 °C
MDL Number: MFCD02093926
CAS Number: 145240-28-4

Order Now

Specification Test Results

Melting point 91-97 ºC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR