4-Methyl-2-(3-pyridinyl)-5-thiazolecarboxylic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Methyl-2-(pyridin-3-yl)thiazole-5-carboxylic acid
034867-1G
101838-524EA
60
USD
101838-524
4-Methyl-2-(3-pyridinyl)-5-thiazolecarboxylic acid ≥97%
4-Methyl-2-(3-pyridinyl)-5-thiazolecarboxylic acid
Hazard: Irritant
Formula:
C₁₀H₈N₂O₂S MW: 220.25 g/mol Melting Pt: 244…248 (dec.) |
MDL Number:
MFCD00171753 CAS Number: 39091-01-5 |
Specification Test Results
Melting point | 244-248(de |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...