[Hydroxy(tosyloxy)iodo]benzene ≥97%

Supplier: Matrix Scientific

Synonyms: Koser ReagentPhenyliodosohydroxy Tosylate

034736-25G
101838-254EA 64.62 USD
101838-254
[Hydroxy(tosyloxy)iodo]benzene ≥97%
[Hydroxy(tosyloxy)iodo]benzene

Hazard: Irritant

Formula: C₁₃H₁₃IO₄S
MW: 392.21 g/mol
Melting Pt: 131…136 °C
MDL Number: MFCD00011547
CAS Number: 27126-76-7

Order Now

Specification Test Results

Melting point 131-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR