4-Bromo-3-methylbenzenesulfonamide ≥97%

Supplier: Matrix Scientific

032360-1G
101833-566EA 60 USD
101833-566
4-Bromo-3-methylbenzenesulfonamide ≥97%
4-Bromo-3-methylbenzenesulphonamide

Hazard: Irritant

Formula: C₇H₈BrNO₂S
Melting Pt: 148 °C
MDL Number: MFCD0394591
CAS Number: 879487-75-9

Order Now

Specification Test Results

Melting point 148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR