3,5-Difluoropicolinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3,5-Difluoropyridine-2-carboxylic acid

031252-5G
101831-314EA 73.85 USD
101831-314
3,5-Difluoropicolinic acid ≥97%
3,5-Difluoropicolinic acid

Hazard: Irritant

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Melting Pt: 202…204 °C
MDL Number: MFCD05664124
CAS Number: 745784-04-7

Order Now

Specification Test Results

Melting point 202-204°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR