1-[(5-Bromo-2-methyl-4-methylsulfonyl)phenyl]piperidine-3-carboxylic acid

Supplier: Matrix Scientific

024979-500MG
101819-330EA 401.54 USD
101819-330
1-[(5-Bromo-2-methyl-4-methylsulfonyl)phenyl]piperidine-3-carboxylic acid
1-[(5-Bromo-2-methyl-4-methylsulfonyl)phenyl]piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₈BrNO₄S
MW: 376.28 g/mol
Melting Pt: 113…115 °C
MDL Number: MFCD09264561
CAS Number: 1000018-36-9

Order Now

Specification Test Results

Melting point 113 - 115

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR