1-[(5-Bromo-2-methyl-4-methylsulfonyl)phenyl]piperazine

Supplier: Matrix Scientific

024978-500MG
101819-328EA 352.31 USD
101819-328
1-[(5-Bromo-2-methyl-4-methylsulfonyl)phenyl]piperazine
1-[(5-Bromo-2-methyl-4-methylsulfonyl)phenyl]piperazine

Hazard: Irritant

Formula: C₁₂H₁₇BrN₂O₂S
Melting Pt: 121…122 °C
MDL Number: MFCD09264560
CAS Number: 1000018-35-8

Order Now

Specification Test Results

Melting point 121 - 122

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR