1-[(5-Bromo-2-methyl-4-methylsulfonyl)phenyl]piperidine-4-carboxamide

Supplier: Matrix Scientific

024977-500MG
101819-326EA 461.54 USD
101819-326
1-[(5-Bromo-2-methyl-4-methylsulfonyl)phenyl]piperidine-4-carboxamide
1-[(5-Bromo-2-methyl-4-methylsulfonyl)phenyl]piperidine-4-carboxamide

Hazard: Irritant

Formula: C₁₄H₁₉BrN₂O₃S
Melting Pt: 296…298 °C
MDL Number: MFCD09264559
CAS Number: 1000018-34-7

Order Now

Specification Test Results

Melting point 296 - 298

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR