5-Bromo-2-methyl-4-methylsulfonylphenylhydrazine

Supplier: Matrix Scientific

024976-500MG
101819-324EA 361.54 USD
101819-324
5-Bromo-2-methyl-4-methylsulfonylphenylhydrazine
5-Bromo-2-methyl-4-methylsulfonylphenylhydrazine

Hazard: Irritant

Formula: C₈H₁₁BrN₂O₂S
Melting Pt: 195…197 °C
MDL Number: MFCD09264558
CAS Number: 1000018-33-6

Order Now

Specification Test Results

Melting point 195 - 197

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR