1-[(4-Methylsulfonyl)naphth-1-yl]piperidine-4-carboxamide

Supplier: Matrix Scientific

024972-250MG
101819-316EA 283.08 USD
101819-316
1-[(4-Methylsulfonyl)naphth-1-yl]piperidine-4-carboxamide
1-[(4-Methylsulfonyl)naphth-1-yl]piperidine-4-carboxamide

Hazard: Irritant

Formula: C₁₇H₂₀N₂O₃S
Melting Pt: 227…230 °C
MDL Number: MFCD09264556
CAS Number: 1000018-32-5

Order Now

Specification Test Results

Melting point 227 - 230o

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR