5-(Hydroxymethyl)-2-methyl-4-(trifluoromethyl)-1,3-thiazole

Supplier: Matrix Scientific

Synonyms: 5-Hydroxymethyl-2-methyl-4-trifluoromethyl-1,3-thiazole

024883-250MG
101819-144EA 315.38 USD
101819-144
5-(Hydroxymethyl)-2-methyl-4-(trifluoromethyl)-1,3-thiazole
5-(Hydroxymethyl)-2-methyl-4-(trifluoromethyl)-1,3-thiazole

Hazard: Irritant

Formula: C₆H₆F₃NOS
MW: 197.178 g/mol
Melting Pt: 83…85 °C
MDL Number: MFCD09264553
CAS Number: 14857-19-3

Order Now

Specification Test Results

Melting point 83 - 85 oC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR