3-[(4-Methylsulfonyl)phenoxy]benzoic acid

Supplier: Matrix Scientific

024880-500MG
101819-138EA 247.69 USD
101819-138
3-[(4-Methylsulfonyl)phenoxy]benzoic acid
3-[(4-Methylsulfonyl)phenoxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₂O₅S
Melting Pt: 160…163 °C
MDL Number: MFCD09264550
CAS Number: 1000018-30-3

Order Now

Specification Test Results

Melting point 160 - 163

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR