1-[(4-Fluoro-2-methylsulfonyl)phenyl]-4-methylpiperazine

Supplier: Matrix Scientific

024876-500MG
101819-130EA 315.38 USD
101819-130
1-[(4-Fluoro-2-methylsulfonyl)phenyl]-4-methylpiperazine
1-[(4-Fluoro-2-methylsulfonyl)phenyl]-4-methylpiperazine

Hazard: Irritant

Formula: C₁₂H₁₇FN₂O₂S
Melting Pt: 97…99 °C
MDL Number: MFCD09264547
CAS Number: 1000339-70-7

Order Now

Specification Test Results

Melting point 97 - 99 oC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR