4-(Benzenesulfonyl)benzaldehyde

Supplier: Matrix Scientific

Synonyms: 4-(Phenylsulfonyl)benzaldehyde4-(Phenylsulphonyl)benzaldehyde

024874-500MG
101819-126EA 186.15 USD
101819-126
4-(Benzenesulfonyl)benzaldehyde
4-(Benzenesulfonyl)benzaldehyde

Hazard: Irritant

Formula: C₁₃H₁₀O₃S
MW: 246.29 g/mol
Melting Pt: 124…126 °C
MDL Number: MFCD00444612
CAS Number: 66-39-7

Order Now

Specification Test Results

Melting point 124 - 126

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR