3-Acetyl-6-chloro-2-methylimidazo[1,2-b]pyridazine

Supplier: Matrix Scientific

024868-500MG
101819-112EA 440 USD
101819-112
3-Acetyl-6-chloro-2-methylimidazo[1,2-b]pyridazine
3-Acetyl-6-chloro-2-methylimidazo[1,2-b]pyridazine

Hazard: Irritant

Formula: C₉H₈ClN₃O
Melting Pt: 127…131 °C (dec.)
MDL Number: MFCD09264542
CAS Number: 90734-73-9

Order Now

Specification Test Results

Melting point 127 - 131

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR