4-Fluoro-1-methylsulfonyl-2-nitrobenzene

Supplier: Matrix Scientific

024861-500MG
101819-100EA 315.38 USD
101819-100
4-Fluoro-1-methylsulfonyl-2-nitrobenzene
4-Fluoro-1-methylsulfonyl-2-nitrobenzene

Hazard: Irritant

Formula: C₇H₆FNO₄S
Melting Pt: 143…145 °C
MDL Number: MFCD09264537
CAS Number: 518990-79-9

Order Now

Specification Test Results

Melting point 143 - 145

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR