2-(Trifluoromethyl)nicotinic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Trifluoromethylnicotinic acid
024843-5G
024843-1G
101819-050EA
64.62
USD
101819-050
101819-048
2-(Trifluoromethyl)nicotinic acid ≥97%
2-(Trifluoromethyl)nicotinic acid
Hazard: Irritant
Formula:
C₇H₄F₃NO₂ MW: 191.11 g/mol Melting Pt: 184…188 °C |
MDL Number:
MFCD01862052 CAS Number: 131747-43-8 |
Specification Test Results
Melting point | 184-188°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...