4-Chlorothieno[3.2.c]pyridine ≥97%

Supplier: Matrix Scientific

024754-1G
101818-826EA 150.77 USD
101818-826
4-Chlorothieno[3.2.c]pyridine ≥97%
4-Chlorothieno[3.2.c]pyridine

Hazard: Irritant

Formula: C₇H₄ClNS
MW: 169.63 g/mol
Melting Pt: 97…100 °C
MDL Number: MFCD01861830
CAS Number: 27685-94-5

Order Now

Specification Test Results

Melting point 97-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR