N-(4-Methoxypyridin-2-yl)-2,2-dimethylpropionamide ≥98%

Supplier: Matrix Scientific

024714-250MG
101818-716EA 303.08 USD
101818-716
N-(4-Methoxypyridin-2-yl)-2,2-dimethylpropionamide ≥98%
N-(4-Methoxypyridin-2-yl)-2,2-dimethylpropionamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MW: 208.26 g/mol
Melting Pt: 82 °C
MDL Number: MFCD08235130
CAS Number: 898561-60-9

Order Now

Specification Test Results

Melting point 82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR