3-(1H-Pyrrolo[2,3-b]pyridin-5-yl)acrylic acid methyl ester ≥99%

Supplier: Matrix Scientific

024689-250MG
101818-646EA 444.62 USD
101818-646
3-(1H-Pyrrolo[2,3-b]pyridin-5-yl)acrylic acid methyl ester ≥99%
3-(1H-Pyrrolo[2,3-b]pyridin-5-yl)acrylic acid methyl ester

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
Melting Pt: 192…193 °C
MDL Number: MFCD09260185
CAS Number: 945029-05-0

Order Now

Specification Test Results

Melting point 192-193°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR