3-Iodo-4-methoxy-1H-pyrrolo[2,3-b]pyridine ≥98%

Supplier: Matrix Scientific

024686-250MG
101818-640EA 183.08 USD
101818-640
3-Iodo-4-methoxy-1H-pyrrolo[2,3-b]pyridine ≥98%
3-Iodo-4-methoxy-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₈H₇IN₂O
Melting Pt: 220…221 °C
MDL Number: MFCD08741528
CAS Number: 928653-75-2

Order Now

Specification Test Results

Melting point 220-221°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR