2-(1H-Pyrazol-3-yl)pyridine ≥95%

Supplier: Matrix Scientific

024679-1G
101818-626EA 44.62 USD
101818-626
2-(1H-Pyrazol-3-yl)pyridine ≥95%
2-(1H-Pyrazol-3-yl)pyridine

Hazard: Irritant

Formula: C₈H₇N₃
MW: 145.16 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD00115151
CAS Number: 75415-03-1

Order Now

Specification Test Results

Melting point 119-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR