4-Bromo-2-nitrophenylhydrazine hydrochloride ≥95%

Supplier: Matrix Scientific

024675-1G
101818-618EA 75.38 USD
101818-618
4-Bromo-2-nitrophenylhydrazine hydrochloride ≥95%
4-Bromo-2-nitrophenylhydrazine hydrochloride

Hazard: Irritant/Store in Freezer

Formula: C₆H₇BrClN₃O₂
Melting Pt: 184…185 °C (dec.)
MDL Number: MFCD09260482
CAS Number: 100032-79-9

Order Now

Specification Test Results

Melting point 184-185°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR