4-Amino-1H-pyrazole ≥97%

Supplier: Matrix Scientific

Synonyms: 1H-Pyrazol-4-amine4-Aminopyrazole

024672-1G
101818-608EA 141.54 USD
101818-608
4-Amino-1H-pyrazole ≥97%
4-Amino-1H-pyrazole

Hazard: Irritant/Refrigerate

Formula: C₃H₅N₃
MW: 83.09 g/mol
Melting Pt: 75…77 °C
MDL Number: MFCD01693729
CAS Number: 28466-26-4

Order Now

Specification Test Results

Melting point 75-77°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR