2-Bromo-3-fluoro-3-methylbutyric acid ≥97%

Supplier: Matrix Scientific

024655-1G
101818-568EA 255.38 USD
101818-568
2-Bromo-3-fluoro-3-methylbutyric acid ≥97%
2-Bromo-3-fluoro-3-methylbutyric acid

Hazard: Irritant

Formula: C₅H₈BrFO₂
Melting Pt: 54…56 °C
MDL Number: MFCD08704576
CAS Number: 50884-94-1

Order Now

Specification Test Results

Melting point 54-56°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR