3-Methoxybenzenesulfonamide ≥97%

Supplier: Matrix Scientific

024638-5G
101818-520EA 149.5 USD
101818-520
3-Methoxybenzenesulfonamide ≥97%
3-Methoxybenzenesulphonamide

Hazard: Irritant

Formula: C₇H₉NO₃S
Melting Pt: 131…133 °C
MDL Number: MFCD08704579
CAS Number: 58734-57-9

Order Now

Specification Test Results

Melting point 131-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR