2,5-Bis(trifluoromethyl)benzenesulfonamide ≥98%

Supplier: Matrix Scientific

024621-1G 024621-5G
101818-466EA 43.08 USD
101818-466 101818-468
2,5-Bis(trifluoromethyl)benzenesulfonamide ≥98%
2,5-Bis(trifluoromethyl)benzenesulphonamide

Hazard: Irritant

Formula: C₈H₅F₆NO₂S
Melting Pt: 145…148 °C
MDL Number: MFCD09475420
CAS Number: 951625-11-9

Order Now

Specification Test Results

Melting point 145-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR