4-Phenoxybenzenesulfonamide ≥98%

Supplier: Matrix Scientific

024620-1G 024620-5G
101818-462EA 331.22 USD
101818-462 101818-464
4-Phenoxybenzenesulfonamide ≥98%
4-Phenoxybenzenesulfonamide

Hazard: Irritant

Formula: C₁₂H₁₁NO₃S
MW: 249.29 g/mol
Melting Pt: 130…132 °C
MDL Number: MFCD00623227
CAS Number: 123045-62-5

Order Now

Specification Test Results

Melting point 130-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR