4-Bromo-2,6-dinitrotoluene ≥97%

Supplier: Matrix Scientific

024588-1G
101818-380EA 95.38 USD
101818-380
4-Bromo-2,6-dinitrotoluene ≥97%
4-Bromo-2,6-dinitrotoluene

Hazard: Irritant

Formula: C₇H₅BrN₂O₄
Melting Pt: 86…88 °C
MDL Number: MFCD07357280
CAS Number: 95192-64-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 86-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR