2,6-Diaminopimelic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 1,5-Diaminoheptanedioic Acid

024587-5G 024587-1G
101818-376EA 100 USD
101818-376 101818-378
2,6-Diaminopimelic acid ≥97%
2,6-Diaminopimelic acid

Hazard: Irritant

Formula: C₇H₁₄N₂O₄
MW: 190.20 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00002637
CAS Number: 583-93-7

Order Now

Specification Test Results

Melting point 300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR