1-[4-(Methylsulfonyl)phenyl]piperidine-3-carboxylic acid

Supplier: Matrix Scientific

024365-250MG
101817-954EA 320 USD
101817-954
1-[4-(Methylsulfonyl)phenyl]piperidine-3-carboxylic acid
1-[4-(Methylsulfonyl)phenyl]piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₄S
Melting Pt: 159…161 °C
MDL Number: MFCD08692418
CAS Number: 942474-19-3

Order Now

Specification Test Results

Melting point 159-161°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR