1H-Indazole-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: Indazole-3-carboxylic acid

027214-25G
101823-746EA 21.54 USD
101823-746
1H-Indazole-3-carboxylic acid ≥97%
1H-Indazole-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 266…270 °C
MDL Number: MFCD00211066
CAS Number: 4498-67-3

Order Now

Specification Test Results

Melting point 266-270°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR