3,4,5-Trifluorobenzenesulfonamide ≥97%
Supplier: Matrix Scientific
026762-5G
026762-1G
101822-818EA
72.31
USD
101822-818
101822-820
3,4,5-Trifluorobenzenesulfonamide ≥97%
3,4,5-Trifluorobenzenesulphonamide
Hazard: Irritant
Formula:
C₆H₄F₃NO₂S MW: 211.16 Melting Pt: 124…126 °C |
MDL Number:
MFCD09800668 CAS Number: 913472-55-6 |
Specification Test Results
Melting point | 124-126°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...