2,4-Dimethylbenzenesulfonamide ≥97%

Supplier: Matrix Scientific

026303-1G
101822-044EA 106.15 USD
101822-044
2,4-Dimethylbenzenesulfonamide ≥97%
2,4-Dimethylbenzenesulfonamide

Hazard: Irritant

Formula: C₈H₁₁NO₂S
MW: 185.25 g/mol
Melting Pt: 136…137 °C
MDL Number: MFCD00460793
CAS Number: 7467-12-1

Order Now

Specification Test Results

Melting point 139-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR