1-Methyl-1H-pyrrolo[2,3-b]pyridin-5-ol

Supplier: Matrix Scientific

026226-100MG
101821-866EA 173.85 USD
101821-866
1-Methyl-1H-pyrrolo[2,3-b]pyridin-5-ol
1-Methyl-1H-pyrrolo[2,3-b]pyridin-5-ol

Hazard: Irritant

Formula: C₈H₈N₂O
Melting Pt: 137.5…137.9 °C
MDL Number: MFCD08741543
CAS Number: 737003-45-1

Order Now

Specification Test Results

Melting point 137.5-137.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR