2-Chloro-5-fluoronicotinic acid ≥95%

Supplier: Matrix Scientific

026180-1G 026180-5G
101821-768EA 21.85 USD
101821-768 101821-766
2-Chloro-5-fluoronicotinic acid ≥95%
2-Chloro-5-fluoronicotinic acid

Hazard: Irritant

Formula: C₆H₃ClFNO₂
MW: 175.5449 g/mol
Melting Pt: 141…142 °C
MDL Number: MFCD03092932
CAS Number: 38186-88-8

Order Now

Specification Test Results

Melting point 141-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR