6-Bromopyridine-2-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromopicolinonitrile6-Bromo-2-pyridinecarbonitrile

020613-1G 020613-5G
101810-448EA 64.62 USD
101810-448 101810-446
6-Bromopyridine-2-carbonitrile ≥95%
6-Bromopyridine-2-carbonitrile

Hazard: Irritant

Formula: C₆H₃BrN₂
MW: 183.0100 g/mol
Melting Pt: 100…104 °C
MDL Number: MFCD00234142
CAS Number: 122918-25-6

Order Now

Specification Test Results

Melting point 100-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR