1-(Trifluoromethylcyclopropyl)benzene ≥97%

Supplier: Matrix Scientific

020611-500MG
101810-440EA 70.77 USD
101810-440
1-(Trifluoromethylcyclopropyl)benzene ≥97%
1-(Trifluoromethylcyclopropyl)benzene

Hazard: Flammable

Formula: C₁₀H₉F₃
MW: 186.18 g/mol
Melting Pt: 106…107 °C/25mm
MDL Number: MFCD08059543
CAS Number: 883547-73-7

Order Now

Specification Test Results

Melting point 106-107°C/2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR