5-Bromo-1H-pyrrolo[2,3-b]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-7-azaindole

020022-5G
101809-252EA 26.15 USD
101809-252
5-Bromo-1H-pyrrolo[2,3-b]pyridine ≥95%
5-Bromo-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.04 g/mol
Melting Pt: 178.0…178.8 °C
MDL Number: MFCD06659677
CAS Number: 183208-35-7

Order Now

Specification Test Results

Melting point 178.0-178.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR