1-(Toluene-4-sulfonyl)-1H-pyrrolo[2,3-b]pyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Tosyl-1H-pyrrolo[2,3-b]pyridine

020020-250MG
101809-246EA 67.69 USD
101809-246
1-(Toluene-4-sulfonyl)-1H-pyrrolo[2,3-b]pyridine ≥98%
1-(Toluene-4-sulfonyl)-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₂S
MW: 272.33 g/mol
Melting Pt: 140.2…141.7 °C
MDL Number: MFCD02083277
CAS Number: 348640-02-8

Order Now

Specification Test Results

Melting point 140.2-141.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR