3-Phenoxybenzylamine hydrochloride

Supplier: Matrix Scientific

Synonyms: (3-Phenoxyphenyl)methylamine hydrochloride

019020-500MG
101807-314EA 121.54 USD
101807-314
3-Phenoxybenzylamine hydrochloride
3-Phenoxybenzylamine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₄ClNO
MW: 235.72 g/mol
Melting Pt: 192…193 °C
MDL Number: MFCD07781045
CAS Number: 376637-85-3

Order Now

Specification Test Results

Melting point 192-193°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR