4-(1H-Pyrazol-4-yl)pyridine ≥98%

Supplier: Matrix Scientific

016109-1G
101804-112EA 330.77 USD
101804-112
4-(1H-Pyrazol-4-yl)pyridine ≥98%
4-(1H-Pyrazol-4-yl)pyridine

Hazard: Irritant

Formula: C₈H₇N₃
MW: 145.1613 g/mol
Melting Pt: 195…198 °C
MDL Number: MFCD00480339
CAS Number: 19959-71-8

Order Now

Specification Test Results

Melting point 195-198°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR