2-Fluoro-4-(methylsulfonyl)aniline ≥98%
Supplier: Matrix Scientific
016098-5G
016098-1G
101804-084EA
29.23
USD
101804-084
101804-086
2-Fluoro-4-(methylsulfonyl)aniline ≥98%
2-Fluoro-4-(methylsulfonyl)aniline
Hazard: Irritant
Formula:
C₇H₈FNO₂S MW: 189.21 g/mol Melting Pt: 145…147 °C |
MDL Number:
MFCD06800756 CAS Number: 832755-13-2 |
Specification Test Results
Melting point | 145-147°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...